Learn More
Bioanalytical Systems Inc EC EPSILON BASIC PLUS SFWR
Supplier: Bioanalytical Systems Inc EF1676
Upgrade, Software; BASi; Epsilon EC basic PLUS; Includes file for upgrading allowed techniques: Sampled Curret Polarography, Normal Pulse Polarography/Voltammetry, Differential Puls Polarography/Voltammetery, etc.