Learn More
Bioanalytical Systems Inc EC BULK ELECTROLYSIS CELL
Supplier: Bioanalytical Systems Inc MF1056
This cell is designed for complete electrolysis of a species in solution, as required for bulk electrolysis and controlled potential coulometry. Ideal for small-scale electrosynthesis (mg quantities).