Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ Eagle Open Head Poly Drum
Eagle Open Head Poly Drum is ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration.
Supplier: Justrite™ 1655BLK
Description
- Constructed of black blow-molded high-density polyethylene with UV inhibitors
- Durable and lightweight
- Weather and chemical resistant
- White lid with a plastic lever-lock band closure
- Lab Packs/Open Head Drums - for use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials - if leaking, must pack with sufficient sorbents for - NO FREE LIQUIDS.
Specifications
DOT 49 CFR 173.12 | |
55 gal. | |
Black | |
Plastic Lever-lock Ring | |
99.4 cm |
Plastic Lever-lock | |
208.2 L | |
HDPE | |
39 1/8 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction