Learn More
SiliCycle™ E-PAK™ Cartridges, SiliaMetS™ Thiol
E-PAK™ Cartridges, SiliaMetS™ Thiol (Si-Thiol) is our most versatile and robust metal scavenger for a variety of metals under a wide range of conditions conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Supplier:  SiliCycle™ LSR51030B5103
 View more versions of this product
Catalog No. 01-182-104
Add to Cart
Description
Description
Metals removed:
  • Best scavenger: Ag, Hg, Os, Pd, and Ru
  • Good scavenger: Cu, Ir, Pb, Rh, Se, and Sn
Additional Information:
  • To facilitate the evaluations of small samples, we can provide loose media and lab scale cartridges on a custom basis.
  • Testing with loose media can be done with samples as small as a few milliliters and is normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
  • Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
  • Lab cartridges can be operated with a peristaltic pump or a small pressure vessel.
Specifications
Specifications
Lab Scale E-PAK Cartridge
White to yellow
3 Pk.
40 to 63 μm
75 g
Lower residual metal concentration in Ag, Hg, Os, Pd, Ru
10 x 5 cm
2 to 9
Thiol
Product Suggestions
Product Suggestions