Learn More
Perkin Elmer US LLC GemCone Low-Flow Nebulizer for Optima 3X00
Supplier:  Perkin Elmer US LLC N0690671
Encompass_Preferred
GemCone Nebulizers require an end cap (N0680504) for use with Scott Spray Chambers.
Catalog No. 50-904-3701