Learn More
AB Sciex LLC eCAP Neutral Capillary 50μm ID 67 cm Total Length (qty 1)
Supplier: AB Sciex LLC 477441
Capillary for use in PA 800 series and P/ACE MDQ Capillary Cartridges for separation and analysis of a variety of samples including pharmaceuticals industrial chemicals chiral drugs nucleic acids amino acids proteins and peptides. This capillary is also used with Advanced cIEF Starter Kit (part number A80976). Capillaries must be trimmed to optimize speed/resolution. Neutral Coated Capillary with laser burned window - 50μm ID x 67 cm total length.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.