Learn More
Dynamic Diagnostics Safety Scraper
Help keep your laboratory environment clear of paraffin and hard-to-remove debris
Supplier: Dynamic Diagnostics 210062
Description
Retractable and use the traditional single edge razor blade
- Consists of a stainless steel blade with a unique locking system to eliminate sliding concerns
- Easily change the blade with a push of the non-slip trigger
Specifications
Safety scraper | |
Steel | |
Unique locking system |