Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynamic Diagnostics Ultra Comfort Manual Phlebotomy Chair

Provides security and comfort for patients, while improving efficiency of operation
Supplier: Dynamic Diagnostics 8675
Description
- Hydraulic foot-pedal height adjustment
- Manual recline
- 360° rotation
- Seat height range of 24 to 32 in. (61 to 81cm)
- Seamless top helps prevent cross-contamination and allows for easy cleaning
- Maximum weight capacity: 250 lb. (113kg)
Specifications
67 x 31.5 x 24 to 32 in. (170.2 x 80 x 61 to 81 cm) | |
24 to 32 in. | |
61 to 81 cm | |
67 in. | |
31.5 in. | |
80 cm | |
250 lb. | |
113 kg |
Provides security and comfort for patients, while improving efficiency of operation | |
38 in. | |
96.5 cm | |
170.2 cm | |
18 in. | |
45.72 cm | |
Hydraulic Phlebotomy Chair |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title