Learn More
Vector Laboratories DyLight 594 Labeled Lycopersicon Esculentum (Tomato) Lectin (LEL,TL) 1 mg
Supplier: Vector Laboratories DL11771
Tomato lectin (from Lycopersicon esculentum) is an effective marker of blood vessels and microglial cells in rodents. Conjugation of the lectin with a fluorophore facilitates fast, one-step detection and visualization using intravascular perfusion methods or direct application to tissue sections. Fluorescent dye-labeled tomato lectin is a valuable tool for studying rodent tumor angiogenesis, tracing neovascular development in xenograft models and brain research. DyLight 594-labeled tomato lectin emits in red and is an excellent second label in systems with green/yellow fluorescence such as GFP expressed in transgenic animals, or with fluorescein conjugates in standard double-label studies. Excitation maximum: 592 nmEmission maximum: 617 nmColor: Red. Inhibiting/Eluting Sugar: Chitin Hydrolysate
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.