Learn More
Vector Laboratories DyLight 488 Labeled Lycopersicon Esculentum (Tomato) Lectin (LEL,TL) 1 mg
Supplier: Vector Laboratories DL11741
Tomato lectin (from Lycopersicon esculentum) is an effective marker of blood vessels and microglial cells in rodents. Conjugation of the lectin with a fluorophore facilitates fast, one-step detection and visualization using intravascular perfusion methods or direct application to tissue sections. Fluorescent dye-labeled tomato lectin is a valuable tool for studying rodent tumor angiogenesis, tracing neovascular development in xenograft models and brain research. DyLight 488-conjugated tomato lectin fluoresces green (emission maximum 518 nm) and has an excitation maximum at 493 nm. Inhibiting/Eluting Sugar: Chitin Hydrolysate
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.