Learn More
Genscript Corporation Antibody Kit Protein Purification Protein Purification 1/EA
Supplier: Genscript Corporation L00455B
DYKDDDDK (Flag) Tag Antibody Plate is a microtiter plate coated with THETM Anti DYKDDDDK (Flag) Tag Antibody. The plate is designed for the rapid capturing of DYKDDDDK (Flag) tagged protein expressed in different systems including E. coli yeast and mammalian extracts and cell culture supernatants. It can be applied for the detection of DYKDDDDK (Flag) tag located at N-terminus C-terminus or internal of the protein. The plate demonstrates high sensitivity and high binding capacity. The capacity and sensitivity varies depending on protein size structure and solution buffer. Generally proteins with lower molecular weight (M.W) can be more sensitive to the DYKDDDDK (Flag) antibody plate than ones with higher M.W. The plate is tolerable with several common used reagents. Avoid using detection reagents that can recognize mouse IgG such as anti mouse IgG antibodies and protein A.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.