Learn More
Electron Microscopy Sciences Dust-Off Xl(DPSXLA) 10 OZ
Supplier: Electron Microscopy Sciences 70838
Contains: Formula S (Difluoroethane) - 100% safe for the ozone. The first safe for the environment disposable pressurized duster. Removes dust and dirt with pure moisture free blasts. There is more product at less cost per ounce. Extension tube is included with the can. Its new concentrated formula will give you 57% more blasts per ounce, 17% more blasts per can.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.