Learn More
Kimberly-Clark Professional™ Dust and Impact Goggle
Click to view available options
Jackson Safety™ Unispec II Safety Eyewear
Description
- Economical impact and particle protection
- Clear lenses allow maximum visible light transmission
- Ideal for indoor use
- Hardcoated lenses for scratch resistance and longer life
- Lenses are made of a polycarbonate material that provides 99.9% UVA/UVB/UVC protection
- Meets ANSI Z87.1+ 2010 standards for high impact with a D4 rating for dust protection
Specifications
Certifications/Compliance | Meets ANSI Z87.1+ 2010 standards for high impact with a D4 rating for dust protection |
Lens Tint | Clear |
Product Type | Safety goggles |
No. per Pack | 200/Pk. |
Ventilation Details | Direct |
Material | Polycarbonate |
UV Protection | 0.999 |