Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences™ DURAN™ YOUTILITY™ GL 45 Bottle Tag
Click to view available options
Color:
Black
Blue
Green
Orange
Purple
Red
White
Yellow
Description
These colorful GL 45 Bottle Tags offer an elegant solution for the marking of laboratory bottles.
- Available in different colors for marking of laboratory bottles and personalization
- Elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
- Feature a graspable tab that makes them easy to remove with bare or gloved hands
- Chemically robust, heat resistant and totally reusable
- Fully compatible with all the GL 45 threads and therefore highly versatile
Specifications
Quantity | 20 Tags/Pk. |
Type | Tag |
Color | White |
Material | Silicone |
For Use With (Application) | Color Coding and Identification |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction