Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Standard Taper Socket Side Arm, Unground Neck, without Stopper
Click to view available options
Capacity (Metric):
5000 mL
Description
Aspirator bottles, ground glass side-arm fro dispensing of liquids. Complying with ISO 4796, these aspirators have a ground socket side-arm and plain neck to accept rubber stoppers. Compatible stopcocks assemblies are available separately.
- Outlet facilitates attachment of ground jointed stopcock
- With plain unground neck for rubber stopper and bottom ground joint outlet
- Made of virtually inert, chemically resistant DURAN™ glass
Specifications
Diameter (Metric) Outer | 182 mm |
Capacity (Metric) | 5000 mL |
Material | Borosilicate Glass (3.3) |
For Use With (Application) | Storage and dispensing of liquids and reagents |
Neck Style | Standard Taper Joint Bottom 29/32 |
No. per Pack | 1/Pk. |
Autoclavable | Autoclavable |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction