Learn More
StatLab OptiCut LP Microtome Blades, 50/pk
SDP
Supplier:  StatLab CUT7223
Encompass_Preferred
Low profile, .012" blade thickness, PTFE coated, 50/pack. OptiCut Blades are precision manufactured to the highest standards for sharpness, consistency, and durability. A proprietary honing and polishing process ensures consistent sharpness and exceptional durability.
Catalog No. NC1204760