Learn More
Adam Equipment Adam Equipment CPWplus 200M Floor Scale, 440lb/200kg Capacity, 0.1lb/50g Readability
Supplier: Adam Equipment CPWPLUS 200M
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Reliable and durable in the field, office, lab or factory, the CPWplus offers compact portability, simple operation, an ample 11.8” x 11.8” platform, and rugged construction. The CPWplus with battery operation and optional carrying case is ideal for concrete testing in the field. All models are well suited for shipping and receiving applications. For food processing or agriculture the stainless steel platform is removable for easy cleaning. The indicator can be mounted where you need it: on the scale, on a wall, or seated on a desk giving total flexibility for the best viewing position. A bi-directional RS-232 interface allows connection to printers or computers. Using standard commands users can request gross, net and tare weights be sent automatically.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.