Learn More
DuPont™ ProClean™ Limited-Use Cleanroom Open-Face Hoods
Click to view available options
Not Specially Cleaned
Description
- White hoods tie at the neck, and cover the shoulders, neck and top and sides of head
- Provide maximum visibility through large facial opening
- Serged seams
- Universal size
- Clean processed hoods are individually packaged in ISO Class 5 cleanroom
- Not specially cleaned hooks are bulk packaged
Specifications
Size | Universal |
Material | Tyvek |
Color | White |
Closure Type | Ties |
Model | PC663S |
Seam Type | Serged |
Quantity | 100/Cs. |
Includes | Ties at neck |