Learn More
DuPont™ ProClean™ Isolation Gowns
Description
Engineered to match specific cleanroom requirements. Ideal for drug manufacturing, electronic parts assembly and many other pharmaceutical and industrial applications. Available in grades compatible with Class M3.5 (Class 100), Class M4.5 (Class 1000), Class M5.5 (Class 10,000) cleanrooms, as defined by Federal Standard 209E. Made of ProShield™ 2, an innovative high-barrier fabric, Pro/Clean gowns are lightweight, exhibit low-particle shedding and are designed to minimize particulate containment. Open-backed isolation gowns feature bound edges, long waist ties, and an adjustable snap closure at the neck. The wrists have tunnelized elastic for a snug, comfortable fit. Available Class A cleaned or unprocessed in a convenient selection of sizes. Packs of 30.
Specifications
Gender | Unisex |
No. of Pockets | None |
Product Type | General Purpose Gown |
Size | Small |
Material | Tyvek |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.