Learn More
CTL Scientific Supply Corp DUOTEST PH 7-10 DISP,PK/3 RL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 903053
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Duotest indicator pH paper 7.0-10.0, pack of 3 rolls in dispensers with color charts, papers with two complimentary indicator zones on one strip. Gradation of scale: 7.0 - 7.3 - 7.6 - 7.9 - 8.2 - 8.5 - 8.8 - 9.1 - 9.4 - 9.7 - 10.0