Learn More
CTL Scientific Supply Corp DUOTEST PH 3.5-6.8 DIS,PK/3 RL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 903033
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Duotest indicator pH paper 3.5-6.8, pack of 3 rolls in dispensers with color charts, papers with two complimentary indicator zones on one strip. Gradation of scale: 3.5 - 3.8 - 4.1 - 4.4 - 4.7 - 5.0 - 5.3 - 5.6 - 5.9 - 6.2 - 6.5 - 6.8