Learn More
CTL Scientific Supply Corp DUOTEST PH 1-12 DISP,PK/3 RL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 903013
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Duotest indicator pH paper 1.0-12.0, pack of 3 rolls in dispensers with color charts, papers with two complimentary indicator zones on one strip. Gradation of scale: 1 - 2 - 3 - 4 - 5 - 6 - 7 - 8 - 9 - 10 - 11 - 12 For extra rolls see Refill 90311