Learn More
Fine Science Tools Dumont #5 Forceps, Straight, 0.1 x 0.06 mm, 11 cm
Supplier: Fine Science Tools 1125130
Style: #5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Dumoxel/ Length: 11cm