Learn More
United States Biological Corporation Dulbecco’s MEM Powder, 10 L
Supplier: United States Biological Corporation D98020110L
Dulbecco’s MEM is the most widely used modification of BME. It contains a 4-fold higher concentration of amino acids and vitamins. Non-essential amino acids and certain essential trace elements were added. The bicarbonate concentration was increased. The standard formula for DMEM is with 1000mg/ml glucose. DMEM was originally developed for the culture of mice embryonic cells. Today, it finds a broad application of serum free culture of normal and transformed mouse and chicken cells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.