Learn More
Sklar Instruments Cardiology Stethoscope, Sklar Dual Head, Navy Blue, 28 in., Stainless Steel / PVC. Sklar
Supplier: Sklar Instruments 061672
Cardiology Stethoscope, Sklar. Dual Head, Navy Blue, 28 in., Stainless Steel / PVC. Sklar's Dual Head Cardiology Stethoscope allows the user to hear high, medium, and low frequency sounds, specifically those identified with the heart such as blood flow, clicks, and murmurs. The cardiology stethoscope has thicker, shorter PVC tubing than a standard stethoscope. This provides a shorter distance for the sounds to travel and decreases interference of curled tubing that can occur with longer stethoscopes. The result is a more accurate identification of what is being heard. The Sklar Dual Head Cardiology Stethoscope features a stainless steel diaphragm and bell to further improve sound transmission. This stethoscope measures 25 inches and is available in a variety of tubing colors.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.