Learn More
Thermo Scientific DUAL RADIO PLSM VIEW WINDOW
Supplier: Thermo Scientific 842312051591
Sleeve for Duo Torch; Provides a spare sealing window for the radial view periscope on duo; systems.; Compatible with all iCAP 6000 Series Duo Spectrometers.