Learn More
Bellco Glass, Inc. The Bellco Sci-ERA Dual Pan Hot Shaker is a slow speed, large orbit shaker with two independently controlled baths for incubation of electrophoretic gels and filter hybridization media. Stringent temperature control with a sensitivity of +/-0.1oC is usef
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bellco Glass, Inc. 774632220
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
The Bellco Sci-ERA Dual Pan Hot Shaker is a slow speed, large orbit shaker with two independently controlled baths for incubation of electrophoretic gels and filter hybridization media. Stringent temperature control with a sensitivity of +/-0.1oC is usef