Learn More
Andwin Scientific Powerful LED Dual Gooseneck Microscope Illuminator Light
Supplier: Andwin Scientific LED11C
This is an LED illuminator with two 22-3/8" long 3-Watt gooseneck lamps that can be bent into various positions. The controller features independent intensity-control for each lamp, allowing for customized lighting-balance. This high-intensity cool lighting LED system is excellent for all applications including biological and electronics applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.