Learn More
Cole-Parmer Fluke Calibration 9009-B Industrial Dual-Block Calibrator
Supplier: Cole-Parmer 1610167
FEATURES
- Temperatures from -15°C to 350°C in one unit
- Two wells in each block for simultaneous comparison calibrations
- Rugged, lightweight, water-resistant enclosure
- Controlled by precision temperature controller
The unit includes two independently controlled temperature blocks. The hot block provides temperatures from 35°C to 350°C, while the cold block covers the range -15·°C to 110°C. Both blocks are controlled by a precision temperature controller.
Available for sale in the United States and Canada only.