Learn More
Dynex™ DSX™ Automated ELISA Processor
Busy clinical and medium-throughput private and industrial labs require advanced automation that can handle virtually every ELISA immunoassay, all in a reliable, cost-effective package. Look no further than the DSX™
Supplier: Dynex™ 65600
Alert:
Appropriate precautions must be taken when working with biohazardous material.
Description
Specifications
Open System | |
ELISA Assays | |
Assay dependent | |
98 samples on-board | |
One year warranty against defects in parts, materials, and workmanship | |
36 in. | |
32 in. | |
42 in. | |
100/240 V auto-switching | |
Revelation DSX™ Software | |
Absorbance | |
244 kg |
Incubator Module | |
(192) 24 x 8 deep-well strips Maximum dilution: 1 to 36,100 | |
Photometric | |
EN 61326-1:2013EN 61326-2-6:2013 IEC/EN 61010-1:2010 Ed.3 IEC/EN 61010-2-010:2014 Ed.3 IEC/EN 61010-2-101 issued 2015/08/14 Ed. 2 CSA C22.2 No. 61010-1:2012 Ed.3 CSA C22.2 No. 61010-2-101:2015 Ed.2 RoHS Compliant | |
405/690 nm | |
91 cm | |
81 cm | |
106 cm | |
50/60 Hz | |
Windows 7/ Windows 10 | |
537 lb. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
DYNEX makes no representations, warranties or performance claims with respect to the performance of Agility for any specific application, including clinical application or for the use of the Agility system with any reagents, assays or other products.