Learn More
Fitness Mart DRY SPIROMETER SPIRALPETTE
Supplier: Fitness Mart 64450
Measures FVC, 1 to 7 liters Hand size, low cost Turn top dial to re-zero Many of our customers use this device to document progress made in rehab and conditioning. This low cost hand-size dry spirometer measures lung capacity (FVC) from 1 to 7 liters. Also used for pulmonary screening. Uses an air-flow powered, precision gear train to indicate air volume in 100 cc increments from 1000 to 7000 cc. It's very easy to use... simply turn the top dial to re-zero. Includes 50 mouthpieces & brief instructions.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.