Learn More
Thermo Scientific™ Dry-Bags™ Half Fraser Broth
Simplify media preparation in food laboratories with this high quality dehydrated medium in a convenient bag format; just open-fill-dispense, saving precious time in the lab.
$1922.00 - $2168.00
Specifications
Color | Clear, yellow |
---|---|
Content And Storage | 10°C to 30°C |
Description | Dry-Bags Half Fraser Broth |
Product Type | Dehydrated Culture Media |
Quantity | 5/Pk. |
Description
Save time in preparing bulk media and diluents for food microbiology testing using Thermo Scientific™ Dry-Bags™ Half Fraser Broth. The irradiated media is supplied in lightweight, transparent plastic bags — just add water and the medium is ready for use. Each bag makes 20L of medium, which can be dispensed through the outlet tube.
- Quick and easy to prepare — just add deionized water through filter sterilization to each Dry-Bag, and they are ready to use in minutes. Pre-sterilized water can also be added under aseptic conditions.
- Light to transport — there is no weighing of media or sterilization required, reducing the burden on media preparation departments and equipment.
- No handling of glassware or weighing of culture media required, no need to add supplementary reagents — empty bags can be disposed of with normal laboratory waste.
- Quality certificates are available for every batch to help reduce the burden of QC testing.
- Dry-Bags are easily adaptable to laboratory workflows and equipment such as automated diluters.
- Dry-Bags are easy to fill and dispense with minimum training.
- Conveniently packed in flat boxes of five or ten, Dry-Bags take up relatively little space compared to prepared media and can be made up as required.
Specifications
Clear, yellow | |
Dry-Bags Half Fraser Broth | |
5/Pk. |
10°C to 30°C | |
Dehydrated Culture Media | |
20L |
Safety and Handling
ShelfLife : 2 Years
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.