Learn More
Thermo Scientific™ Dry-Bags™ Buffered Peptone Water (ISO)
Simplify media preparation in food laboratories with this high quality dehydrated medium in a convienient bag format; just open-fill-dispense, saving precious time in the lab.
$1898.00 - $2614.00
Specifications
Color | Clear, colorless - straw |
---|---|
Content And Storage | 10°C to 30°C |
Description | Dry-Bags Buffered Peptone Water (ISO) |
Product Type | Pre-enrichment Medium |
Quantity | 10/Pk. |
Catalog Number | Mfr. No. | Format | Yield | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Yield | Price | Quantity & Availability | ||||
DB1049M
|
Thermo Scientific™
DB1049M |
Dry-Bags™, with filter | 20L |
Each for $2,614.00
|
|
||||
DB1049W
|
Thermo Scientific™
DB1049W |
Dry-Bags™, without filter | 20L |
Pack of 10 for $1,898.00
|
|
||||
Description
Save time in preparing bulk media and diluents for food microbiology testing using Thermo Scientific™ Dry-Bags™ Buffered Peptone Water (ISO). The irradiated media is supplied in lightweight, transparent plastic bags — just add water and the medium is ready for use. Each bag makes 20L of medium, which can be dispensed through the outlet tube.
Use Buffered Peptone Water (ISO) as a non-selective pre-enrichment medium for the isolation of Salmonella species from food and associated samples. As Salmonella may be present in low numbers and/or sub-lethally injured, pre-enrichment allows the cells time to repair and multiply before being introduced to selective culture, thereby improving the chances of recovery from the sample.- Quick and easy to prepare — just add deionized water through filter sterilization to each Dry-Bag, and they are ready to use in minutes. Pre-sterilized water can also be added under aseptic conditions.
- Light to transport — there is no weighing of media or sterilization required, reducing the burden on media preparation departments and equipment.
- No handling of glassware or weighing of culture media required, no need to add supplementary reagents — empty bags can be disposed of with normal laboratory waste.
- Quality certificates are available for every batch to help reduce the burden of QC testing.
- Dry-Bags are easily adaptable to laboratory workflows and equipment such as automated diluters.
- Dry-Bags are easy to fill and dispense with minimum training.
- Conveniently packed in flat boxes of five or ten, Dry-Bags take up relatively little space compared to prepared media and can be made up as required.
Specifications
Clear, colorless - straw | |
Dry-Bags Buffered Peptone Water (ISO) | |
10/Pk. |
10°C to 30°C | |
Pre-enrichment Medium | |
20L |
Safety and Handling
ShelfLife : 2 Years
Your input is important to us. Please complete this form to provide feedback related to the content on this product.