Learn More
AVANTOR MASTERFLEX (VWR LLC) Hand-operated drum pump, 316 SS body, 16 strokes/gallon
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0707920
Hand-operated drum pump, 316 SS body, 16 strokes/gallon, Ideal for most organic phase applications, Steel hand-operated drum pumps feature a convenient 1?2" OD spout.To safely pump petroleum-based solvents, use grounding wire (order separately see "Accessories"