Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) DrumQuik Drum Adapter System Kit, 902mm tube and 3/4" barbed connection
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX3160811
The DrumQuik PRO system offers a simplified solution to your chemical dispensing needs. This system maintains a closed system, protecting your facility, your operating environment, and your employees from the potential dangers of chemical drips, spills and escaping fumes. The simple connections, rugged construction, and chemical-resistant materials handle a wide variety of applications—including acids and caustics, agricultural liquids, cleaning agents and sterilants, paints and coatings, and food oils and syrups. The base components are a reusable quick-disconnect coupler and a single-use drum insert assembly; both of which are made from environmentally friendly, food-grade polyethylene (PE). Order a convenient dispensing kit which includes all required components, or customize your own system with individual components. The coupler body and plugs mate equally well with all drum inserts listed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.