Learn More
LabScientific Drosophila agar 80-100 mesh, 1 kg, powder, low gelling temp & high gel strength.
Supplier: LabScientific FLY80201
Drosophila agar 80-100 mesh, 1 kg. Powder. A High Quality Microbiological Agar ~ primarily used in labs as a culture medium in Petri dishes. Gelidium agar has a low gelling temperature (34 to 36 degrees Celsius). It doesn't contain trace metals, soluble carbohydrates, carbohydrates or any spores that would inhibit bacterial growth, making it an exceptional medium for research projects. Gelidium is extracted from the species of red algae Rhodophyceae type form Gelidium Genera. Gelidium is soluble in boiling water, Kosher Certified, and classified as GRAS by the FDA. Melting point 88 C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.