Learn More
United States Biological Corporation DROP OUT MIX - LEUCINE 100G
Supplier: United States Biological Corporation D9525100G
Drop-out Mix Complete (DOC) is used as a supplement to Drop-out Base for making synthetic, complete media formulations in accordance with Methods in Yeast Genetics. All lots are quality control tested using the appropriate yeast strains and transformants. Convenient Powders Useful for One-Hybrid and Two-Hybrid Systems Competitively Priced...! Dust-Free: Specially milled powder blends are designed to eliminate or greatly reduce airborne particulates Homogeneous: Proprietary blending technology ensures complete mixing of components Companion Products: D9500: Drop-out Base With Glucose (Powder) D9501: Drop-out Base With Raffinose (Powder) D9502: Drop-out Base With Galactose (Powder) D9510: Drop-out Base With Glucose and Agar (Powder) D9511: Drop-out Base With Raffinose and Agar (Powder) D9512: Drop-out Base With Galactose and Agar (Powder) Y2020: Yeast Nitrogen Base w/ AA & w/ AS, w/o Carbohydrate (YNB) (Powder)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.