Learn More
Perkin Elmer US LLC System 2 EDL Driver for AAnalyst
Supplier: Perkin Elmer US LLC 03030997
Includes a short (2.1 ft) cable. If a longer length is needed, an extension cable is available (03030998). An adapter (N3150303) is required for use on AAnalyst™ 200/400 series instruments.