Learn More
New Pig Drive-Over DrainBlocker™ Drain Cover
Looking for a tough storm-drain cover? You can drive tankers over this one.
Supplier: New Pig PLR903
This item is not returnable.
View return policy
Description
- Confidently block drains from spills and water with this outdoor drain cover's super-sealing bottom layer that conforms perfectly for a tight seal while the HogHyde rubber top layer withstands years and years of repeated heavy vehicle traffic
- Trust this drive-over drain cover to resist everything from punctures, stretching and tearing to prolonged exposure to weather, UV/IR light and oils like diesel fuel and gasoline, plus many chemicals
- Maintain a professional look for longer since this drive-over drain cover is designed to wipe clean and get back on the job
- Use it how you need it, either leaving it down as a preventive measure in leak- and spill-prone areas or storing it for emergency spill response
- Respond faster to spills by conveniently carrying the cover in the included nylon storage bag.
Specifications
Drain Cover |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov