Learn More
MSC Value Collection 01241058 1.05mm 118° High Speed Steel Jobber Drill Oxide Finish, Right Hand Cut, Spiral Flute, Straight Shank, 1-39/64" Overall Length, Standard Point
Supplier: MSC 01241058
Value Collection 01241058 1.05mm 118° High Speed Steel Jobber Drill Oxide Finish, Right Hand Cut, Spiral Flute, Straight Shank, 1-39/64" Overall Length, Standard Point