Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Safety Apparel™ DRIFIRE™ Performance FR Jean 34 in.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
6X-Large
Description
- CAT 2
- Fabric/Material: 14.75 oz. High Performance Denim
- Dual certified to protect against flash fire and arc flash hazards
- Ultimate heavy duty performance utility pant
- Breathable fabric which increases moisture management
- 5 pocket style and classic cut
- Double tool pocket on the right side
- Inherently flame resistant
- FR protection will not wash/wear out
- Quickly identify compliance with exterior labeling
- Dries up to 37% faster than competitors
- Leg openings made to fit over work boots
- Comfort washed
- Made in the USA
Specifications
Size | 6X-Large |
Material | Denim |
Color | Blue |
No. of Pockets | 4 |
Arc Flash Rating | 15 cal/cm2 |
Certifications/Compliance | NFPA 70E for Arc Flash, ASTM F1506 for Arc Flash, CSA Z462 for Arc Flash (Canada), OSHA 1910.269 |
Size (English) Inseam | 34 in. |
Size (English) Waist | 48 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction