Learn More
Thermo Scientific™ DRI™ Negative Urine Drugs of Abuse Calibrators
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation.
$273.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | DRI Negative Urine Drugs of Abuse Calibrators |
Concentration | 0 ng/mL |
Detectable Analytes | Negative Calibrator |
Sample Type | Urine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-401
|
Thermo Scientific™
1388 |
1 x 25 mL |
Each for $327.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-422
|
Thermo Scientific™
1664 |
1 x 10 mL |
Each for $273.00
|
|
|||||
Description
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Negative Urine Drugs of Abuse Calibrators — ready-to-use assays in liquid form. These assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Applications for an array of clinical chemistry analyzers
- Variety of calibrators and multiconstituent control sets
Specifications
2°C to 8°C | |
0 ng/mL | |
Urine | |
1 Bottle |
DRI Negative Urine Drugs of Abuse Calibrators | |
Negative Calibrator | |
Calibrator |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.