Learn More
Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Assay
Save time while maintaining accuracy with the liquid, ready-to-use Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Assay.
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | DRI Hydrocodone Control Set |
Detectable Analytes | Hydrocodone |
DoA Calibrators | DRI Hydrocodone Calibrators, DRI Negative Urine Calibrators |
Sample Type | Urine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-376
|
Thermo Scientific™
10018054 |
3 x 18 mL |
Each of 1 for $2,745.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of the Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Assay, which offers applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs. This assay is for sale in the U.S.A. and Canada only. Supports new SAMHSA guidelines (effective October 1, 2017)
- Supports new SAMHSA guidelines (effective October 1, 2017)
- Specific detection of hydrocodone
- Qualitative or semi-quantitative modes
- Excellent correlation to LC-MS/MS
- Applications for an array of clinical chemistry analyzers
- For sale in the U.S.A. and Canada only
Specifications
2°C to 8°C | |
Hydrocodone | |
Urine |
DRI Hydrocodone Control Set | |
DRI Hydrocodone Calibrators, DRI Negative Urine Calibrators | |
Reagent |
Disclaimers: Some of these products are not cleared in every country. Please contact your sales representative for more information. Product claims may differ from country to country based on regulations and product approvals. Please contact your country representative for further details.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.