Learn More
Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Controls
Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Controls.
Specifications
Content And Storage | Room Temperature |
---|---|
Description | Gravity-Detect™ |
Detectable Analytes | Specific Gravity |
Sample Type | Urine |
Type | Control |
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
23-278-425
|
Thermo Scientific™
1756 |
1.015 μg/mL |
Each for $281.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-426
|
Thermo Scientific™
1757 |
1.030 μg/mL |
Each for $281.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Controls. Liquid and ready-to-use, these controls can be performed on a variety of general chemistry analyzers. The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution.
Our adulteration assays provide a complete and automated way to test for sample integrity and adulteration.- Associated calibrators and controls offered in convenient sizes
- Liquid, ready-to-use
- Can be performed on a variety of general chemistry analyzers
Specifications
Room Temperature | |
Specific Gravity | |
Control |
Gravity-Detect™ | |
Urine | |
1 x 25 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.