Learn More
Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators
Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators.
Specifications
Description | DRI™ Gravity-Detect™ Specimen Validity Test Calibrators |
---|---|
Detectable Analytes | Specific Gravity |
Sample Type | Urine |
Type | Calibrator |
Quantity | 1 x 25 mL |
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
23-278-423
|
Thermo Scientific™
1754 |
1.010 |
Each for $267.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-424
|
Thermo Scientific™
1755 |
1.025 |
Each for $267.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators. Liquid and ready-to-use, these controls can be used with the Thermo Scientific™ DRI™ Gravity-Detect™ Reagent Kit on a variety of general chemistry analyzers. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution.
- Can be performed on a variety of general chemistry analyzers
Specifications
DRI™ Gravity-Detect™ Specimen Validity Test Calibrators | |
Urine | |
1 x 25 mL |
Specific Gravity | |
Calibrator |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.