Learn More
Thermo Scientific™ DRI™ Fentanyl Drugs of Abuse Assay
Save time while maintaining accuracy with these liquid, ready-to-use assays, which are for Criminal Justice and Forensic use only and are for sale in the U.S.A. only.
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | DRI Fentanyl Control Sets |
Detectable Analytes | Fentanyl (FEN) |
DoA Calibrators | DRI Fentanyl Calibrators |
Sample Type | Urine |
Alert:
For Criminal Justice and Forensic use only.
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-369
|
Thermo Scientific™
10016006 |
3 x 18 mL |
Each of 1 for $2,710.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Fentanyl Drugs of Abuse Assays. Field-tested on a wide range of clinical analyzers, including the Thermo Scientific™ Indiko™ Clinical Analyzer, these liquid, ready-to-use assays have been widely accepted in laboratories where cost control and optimal productivity are key issues. These assays are for Criminal Justice and Forensic use only and are for sale in the U.S.A. only.
- Specific detection of fentanyl
- Qualitative or semi-quantitative modes
- Excellent correlation to LC-MS/MS
- Applications for an array of clinical chemistry analyzers
Specifications
2°C to 8°C | |
Fentanyl (FEN) | |
Urine |
DRI Fentanyl Control Sets | |
DRI Fentanyl Calibrators | |
Reagent |
For sale in the U.S.A. only.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.