Learn More
Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Assays
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation.
$745.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | DRI Ethyl Alcohol Control Set |
Detectable Analytes | Alcohol |
DoA Calibrators | DRI Ethyl Alcohol Calibrators |
Sample Type | Plasma, Serum, Urine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-300
|
Thermo Scientific™
0037 |
1 x 100 mL |
Each for $2,476.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-374
|
Thermo Scientific™
10016397 |
3 x 18 mL |
Each for $745.00
|
|
|||||
Description
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. These liquid, ready-to-use assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
2°C to 8°C | |
Alcohol | |
Plasma, Serum, Urine |
DRI Ethyl Alcohol Control Set | |
DRI Ethyl Alcohol Calibrators | |
Reagent |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.