Learn More
Apexbio Technology LLC DRB,50mg CAS# 53-85-0
SDP
Supplier:  Apexbio Technology LLC C479850
Encompass_Preferred
5, 6-dichloro-1-β-D-ribofuranosylbenzimidazole (DRB) is a transcriptional elongation inhibitor.Cyclin-dependent kinases (CDKs) belong to a family of protein kinases involved in regulating the cell cycle, transcription, mRNA processing, and the differentiation of nerve cells.[1] They are expressed in all known eukaryotes. With cyclin, CDK shows kinase activity. CDKs are serine-threonine kinases [1].DRB inhibited the activity of several carboxyl-terminal domain (CTD) kinases including casein kinase II, Cdk7, Cdk8, and Cdk9 with the IC50 of 4-10 μM, ~20 μM, ~20 μM, and 3 μM) [2-5]. In HeLa cells, DRB (75 μM) inhibited 60-75% of nuclear heterogeneous RNA (hnRNA) synthesis. DRB (75 μM) reduced the appearance of labeled cytoplasmic poly(A)-containing messenger RNA (mRNA) by approximately 95%. DRB inhibited the initiation of hnRNA chains, but did not directly interfere with labeling of poly(A) [6]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0093