Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ DRAQ7™
$191.70 - $362.00
Specifications
Content And Storage | Store at 4°C in the dark. Do not freeze. |
---|---|
For Use With (Application) | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy |
Specifications
Store at 4°C in the dark. Do not freeze. | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title