Learn More
Mystaire™ Downflow Workstation Particulate Pre-Filter
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For use with the Mystaire Downflow Workstation (FE-WS6). An electrostatically charged fiber filter which effectively removes submicron particulates and is designed to protect the adsorption filter from contaminants such as smoke, acid mists, and other submicron particulates.
Supplier: Mystaire™ FE1013
Description
- For use with ductless downflow workstation
Specifications
Particulate Prefilters | |
32 in. | |
28 in. |
Prefilter | |
80 cm | |
70 cm |